Assay Details
| Gene Symbol: CHR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123848.4 | NP_199293.3 | 9 - 10 | 4987 | 127 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CHR4| Gene Symbol: CHR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123848.4 | NP_199293.3 | 9 - 10 | 4987 | 127 | |
Set Membership: |
|