Assay Details
| Gene Symbol: PP2-A7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344585.1 | NP_001330203.1 | 1 - 2 | 440 | 69 | |
| NM_123877.2 | NP_199322.1 | 2 - 3 | 417 | 69 | ||
More Information
Set Membership: |
|

PP2-A7
Genomic Map