Assay Details
| Gene Symbol: PP2-A7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344585.1 | NP_001330203.1 | 2 - 3 | 642 | 99 | |
| NM_123877.2 | NP_199322.1 | 3 - 4 | 619 | 99 | ||
More Information
Set Membership: |
|

PP2-A7
Genomic Map