Assay Details
| Gene Symbol: RHA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123881.4 | NP_199326.1 | 1 - 2 | 363 | 71 | |
| GenBank mRNA | AF389298.1 | - | 1 - 2 | 226 | 71 | |
| AY097362.1 | - | 1 - 2 | 39 | 71 | ||
| X59152.1 | - | 99 | 71 | |||
More Information
Set Membership: |
|

RHA1
Genomic Map