Assay Details
| Gene Symbol: RHA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344589.1 | NP_001330210.1 | 3 - 4 | 712 | 78 | |
| NM_123881.4 | NP_199326.1 | 3 - 4 | 606 | 78 | ||
| GenBank mRNA | AF389298.1 | - | 3 - 4 | 469 | 78 | |
| AY097362.1 | - | 3 - 4 | 282 | 78 | ||
| X59152.1 | - | 342 | 78 | |||
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length between 71 and 85  Detect Genomic DNA  Arabidopsis thaliana  Probe spans exons | 
Gene Ontology Categories:
 Back To Top
 Back To Top
 RHA1
RHA1 Genomic Map
Genomic Map Back To Top
 Back To Top