Assay Details
| Gene Symbol: RHA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344589.1 | NP_001330210.1 | 5 - 6 | 842 | 112 | |
| NM_123881.4 | NP_199326.1 | 5 - 6 | 736 | 112 | ||
| GenBank mRNA | AF389298.1 | - | 5 - 6 | 599 | 112 | |
| AY097362.1 | - | 5 - 6 | 412 | 112 | ||
More Information
Set Membership: |
|

RHA1
Genomic Map