Assay Details
| Gene Symbol: NRPC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344590.1 | NP_001332060.1 | 27 - 28 | 2509 | 79 | |
| NM_001344591.1 | NP_001332059.1 | 27 - 28 | 2427 | 79 | ||
| NM_123882.5 | NP_199327.4 | 27 - 28 | 2427 | 79 | ||
More Information
Set Membership: |
|

NRPC2
Genomic Map