Assay Details
| Gene Symbol: BMY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036937.1 | NP_001032014.1 | 3 - 4 | 810 | 98 | |
| NM_001344606.1 | NP_001330613.1 | 3 - 4 | 899 | 98 | ||
| NM_123898.3 | NP_199343.1 | 3 - 4 | 814 | 98 | ||
| GenBank mRNA | AK117140.1 | - | 3 - 4 | 811 | 98 | |
| BT006482.1 | - | 3 - 4 | 788 | 98 | ||
More Information
Set Membership: |
|

BMY2
Genomic Map