Assay Details
| Gene Symbol: BMY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036937.1 | NP_001032014.1 | 5 - 6 | 1207 | 72 | |
| NM_001344606.1 | NP_001330613.1 | 5 - 6 | 1296 | 72 | ||
| NM_123898.3 | NP_199343.1 | 5 - 6 | 1211 | 72 | ||
| GenBank mRNA | AK117140.1 | - | 5 - 6 | 1208 | 72 | |
| BT006482.1 | - | 5 - 6 | 1185 | 72 | ||
More Information
Set Membership: |
|

BMY2
Genomic Map