Assay Details
Gene Symbol: GAS41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203546.1 | NP_001190475.1 | 4 - 5 | 707 | 116 | |
NM_123928.4 | NP_199373.1 | 4 - 5 | 862 | 116 | ||
GenBank mRNA | AY072123.1 | - | 4 - 5 | 758 | 116 | |
AY096486.1 | - | 4 - 5 | 601 | 116 | ||
AY463617.1 | - | 4 - 5 | 601 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
