Assay Details
| Gene Symbol: SAG12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123957.3 | NP_568651.1 | 1 - 2 | 519 | 64 | |
| GenBank mRNA | AF370131.1 | - | 1 - 2 | 497 | 64 | |
| AY040073.1 | - | 1 - 2 | 458 | 64 | ||
More Information
Set Membership: |
|

SAG12
Genomic Map