Assay Details
| Gene Symbol: VAM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124057.4 | NP_568671.1 | 5 - 6 | 949 | 69 | |
| GenBank mRNA | AF385695.1 | - | 5 - 6 | 784 | 69 | |
| AY072015.1 | - | 5 - 6 | 790 | 69 | ||
| AY085121.1 | - | 5 - 6 | 806 | 69 | ||
| AY133666.1 | - | 5 - 6 | 634 | 69 | ||
| U88045.1 | - | 5 - 6 | 770 | 69 | ||
More Information
Set Membership: |
|

VAM3
Genomic Map