Assay Details
Gene Symbol: HB-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344703.1 | NP_001318750.1 | 3 - 4 | 436 | 67 | |
NM_001344704.1 | NP_001330010.1 | 3 - 4 | 514 | 67 | ||
NM_124059.4 | NP_199499.3 | 2 - 3 | 572 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
