Assay Details
| Gene Symbol: HB-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344703.1 | NP_001318750.1 | 8 - 9 | 1757 | 76 | |
| NM_001344704.1 | NP_001330010.1 | 8 - 9 | 1835 | 76 | ||
| NM_124059.4 | NP_199499.3 | 7 - 8 | 1893 | 76 | ||
More Information
Set Membership: |
|

HB-7
Genomic Map