Assay Details
| Gene Symbol: LBA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124072.3 | NP_199512.2 | 23 - 24 | 3106 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LBA1| Gene Symbol: LBA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124072.3 | NP_199512.2 | 23 - 24 | 3106 | 69 | |
Set Membership: |
|