Assay Details
Gene Symbol: ERF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124093.3 | NP_199533.1 | 1 | 985 | 122 | |
GenBank mRNA | AB008104.1 | - | 1 | 834 | 122 | |
AK175151.1 | - | 1 | 834 | 122 | ||
AK175229.1 | - | 1 | 848 | 122 | ||
AK176532.1 | - | 1 | 848 | 122 | ||
AK176606.1 | - | 1 | 842 | 122 | ||
AK176825.1 | - | 1 | 848 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
