Assay Details
| Gene Symbol: NUDT8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344713.1 | NP_001331621.1 | 3 - 4 | 749 | 146 | |
| NM_124095.4 | NP_568680.1 | 3 - 4 | 701 | 146 | ||
More Information
Set Membership: |
|

NUDT8
Genomic Map