Assay Details
| Gene Symbol: NUDT8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344713.1 | NP_001331621.1 | 4 - 5 | 895 | 88 | |
| NM_124095.4 | NP_568680.1 | 4 - 5 | 847 | 88 | ||
More Information
Set Membership: |
|

NUDT8
Genomic Map