Assay Details
Gene Symbol: HAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124108.3 | NP_199548.1 | 1 - 2 | 506 | 82 | |
GenBank mRNA | AB067629.1 | - | 1 - 2 | 146 | 82 | |
AF428450.1 | - | 1 - 2 | 408 | 82 | ||
AJ431183.1 | - | 1 - 2 | 146 | 82 | ||
AY052324.1 | - | 1 - 2 | 319 | 82 | ||
AY061904.1 | - | 1 - 2 | 146 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
