Assay Details
| Gene Symbol: NUDT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344736.1 | NP_001331651.1 | 3 - 4 | 888 | 73 | |
| NM_001344737.1 | NP_001331652.1 | 4 - 5 | 569 | 73 | ||
| NM_001344738.1 | NP_001331649.1 | 3 - 4 | 515 | 73 | ||
| NM_001344739.1 | NP_001331650.1 | 3 - 4 | 448 | 73 | ||
| NM_124139.4 | NP_568687.1 | 4 - 5 | 619 | 73 | ||
| GenBank mRNA | AY042806.1 | - | 4 - 5 | 453 | 73 | |
| AY064646.1 | - | 3 - 4 | 377 | 73 | ||
More Information
Set Membership: |
|

NUDT2
Genomic Map