Assay Details
| Gene Symbol: ELP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161296.1 | NP_001154768.1 | 15 - 16 | 2205 | 69 | |
| NM_001344757.1 | NP_001318757.1 | 16 - 17 | 2246 | 69 | ||
| NM_124183.4 | NP_199620.4 | 15 - 16 | 2071 | 69 | ||
More Information
Set Membership: |
|

ELP1
Genomic Map