Assay Details
| Gene Symbol: ELP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161296.1 | NP_001154768.1 | 5 - 6 | 959 | 95 | |
| NM_001344757.1 | NP_001318757.1 | 6 - 7 | 1000 | 95 | ||
| NM_124183.4 | NP_199620.4 | 5 - 6 | 825 | 95 | ||
More Information
Set Membership: |
|

ELP1
Genomic Map