Assay Details
| Gene Symbol: ELP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161296.1 | NP_001154768.1 | 9 - 10 | 1650 | 151 | |
| NM_001344757.1 | NP_001318757.1 | 10 - 11 | 1691 | 151 | ||
| NM_124183.4 | NP_199620.4 | 9 - 10 | 1516 | 151 | ||
More Information
Set Membership: |
|

ELP1
Genomic Map