Assay Details
| Gene Symbol: ELP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161296.1 | NP_001154768.1 | 11 - 12 | 1900 | 71 | |
| NM_001344757.1 | NP_001318757.1 | 12 - 13 | 1941 | 71 | ||
| NM_124183.4 | NP_199620.4 | 11 - 12 | 1766 | 71 | ||
More Information
Set Membership: |
|

ELP1
Genomic Map