Assay Details
| Gene Symbol: FKBP15-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124234.4 | NP_199669.1 | 3 - 4 | 577 | 73 | |
| GenBank mRNA | AF370159.1 | - | 2 - 3 | 135 | 73 | |
| AY059146.1 | - | 2 - 3 | 117 | 73 | ||
| AY085294.1 | - | 2 - 3 | 138 | 73 | ||
More Information
Set Membership: |
|

FKBP15-2
Genomic Map