Assay Details
Gene Symbol: FKBP15-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124234.4 | NP_199669.1 | 4 - 5 | 634 | 94 | |
GenBank mRNA | AF370159.1 | - | 3 - 4 | 192 | 94 | |
AY059146.1 | - | 3 - 4 | 174 | 94 | ||
AY085294.1 | - | 3 - 4 | 195 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
