Assay Details
Gene Symbol: PANC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124261.4 | NP_199695.1 | 3 - 4 | 1120 | 138 | |
GenBank mRNA | AK175238.1 | - | 3 - 4 | 833 | 138 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
