Assay Details
Gene Symbol: SAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124264.4 | NP_199698.1 | 2 - 3 | 220 | 100 | |
GenBank mRNA | AY034896.1 | - | 2 - 3 | 159 | 100 | |
BT025700.1 | - | 2 - 3 | 159 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
