Assay Details
Gene Symbol: SUS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344824.1 | NP_001331823.1 | 2 - 3 | 397 | 136 | |
NM_001344825.1 | NP_001331822.1 | 2 - 3 | 309 | 136 | ||
NM_124296.3 | NP_199730.1 | 3 - 4 | 774 | 136 | ||
GenBank mRNA | KJ138646.1 | - | 3 - 4 | 375 | 136 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
