Assay Details
Gene Symbol: HRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344827.1 | NP_001332206.1 | 1 - 2 | 291 | 75 | |
NM_124300.4 | NP_199734.2 | 1 - 2 | 341 | 75 | ||
GenBank mRNA | AF372963.1 | - | 1 - 2 | 140 | 75 | |
AY133584.1 | - | 1 - 2 | 82 | 75 | ||
HQ322385.1 | - | 1 - 2 | 82 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
