Assay Details
| Gene Symbol: HRB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344827.1 | NP_001332206.1 | 4 - 5 | 721 | 82 | |
| NM_124300.4 | NP_199734.2 | 4 - 5 | 786 | 82 | ||
| GenBank mRNA | AF372963.1 | - | 4 - 5 | 585 | 82 | |
| AY133584.1 | - | 4 - 5 | 527 | 82 | ||
| HQ322385.1 | - | 4 - 5 | 512 | 82 | ||
More Information
Set Membership: |
|

HRB1
Genomic Map