Assay Details
Gene Symbol: PNG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124335.3 | NP_199768.1 | 2 - 3 | 279 | 74 | |
GenBank mRNA | AK228156.1 | - | 2 - 3 | 199 | 74 | |
AM745095.1 | - | 2 - 3 | 135 | 74 | ||
AY140065.1 | - | 2 - 3 | 195 | 74 | ||
BT003398.1 | - | 2 - 3 | 135 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
