Assay Details
Gene Symbol: PNG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124335.3 | NP_199768.1 | 6 - 7 | 701 | 68 | |
GenBank mRNA | AK228156.1 | - | 6 - 7 | 621 | 68 | |
AM745095.1 | - | 6 - 7 | 557 | 68 | ||
AY140065.1 | - | 6 - 7 | 617 | 68 | ||
BT003398.1 | - | 6 - 7 | 557 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
