Assay Details
| Gene Symbol: AT5G50310 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124410.4 | NP_568723.1 | 17 - 18 | 1853 | 113 | |
| GenBank mRNA | AF412054.1 | - | 17 - 18 | 1823 | 113 | |
More Information
Set Membership: |
|

AT5G50310
Genomic Map