Assay Details
| Gene Symbol: AT5G50310 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124410.4 | NP_568723.1 | 3 - 4 | 394 | 138 | |
| GenBank mRNA | AF412054.1 | - | 3 - 4 | 364 | 138 | |
More Information
Set Membership: |
|

AT5G50310
Genomic Map