Assay Details
| Gene Symbol: ELO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124412.3 | NP_568725.1 | 8 - 9 | 1675 | 76 | |
| GenBank mRNA | AJ964958.1 | - | 8 - 9 | 1541 | 76 | |
| AY056323.1 | - | 8 - 9 | 1569 | 76 | ||
More Information
Set Membership: |
|

ELO3
Genomic Map