Assay Details
Gene Symbol: NF-YC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124429.1 | NP_199858.1 | 1 | 568 | 108 | |
GenBank mRNA | DQ056712.1 | - | 1 | 568 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
