Assay Details
| Gene Symbol: UBC27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124465.6 | NP_199900.1 | 3 - 4 | 618 | 79 | |
| GenBank mRNA | AY065303.1 | - | 3 - 4 | 468 | 79 | |
| AY096429.1 | - | 3 - 4 | 390 | 79 | ||
| DQ027040.1 | - | 3 - 4 | 390 | 79 | ||
| DQ677667.1 | - | 3 - 4 | 447 | 79 | ||
More Information
Set Membership: |
|

UBC27
Genomic Map