Assay Details
| Gene Symbol: UBC27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203579.1 | NP_001190508.1 | 4 - 5 | 713 | 78 | |
| NM_124465.6 | NP_199900.1 | 4 - 5 | 700 | 78 | ||
| GenBank mRNA | AY065303.1 | - | 4 - 5 | 550 | 78 | |
| AY096429.1 | - | 4 - 5 | 472 | 78 | ||
| DQ027040.1 | - | 4 - 5 | 472 | 78 | ||
| DQ677667.1 | - | 4 - 5 | 529 | 78 | ||
More Information
Set Membership: |
|

UBC27
Genomic Map