Assay Details
| Gene Symbol: RHD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124485.3 | NP_199919.1 | 7 - 8 | 2190 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RHD2| Gene Symbol: RHD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124485.3 | NP_199919.1 | 7 - 8 | 2190 | 68 | |
Set Membership: |
|