Assay Details
Gene Symbol: ERD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124486.3 | NP_568750.1 | 2 - 3 | 1186 | 69 | |
GenBank mRNA | AY035112.1 | - | 2 - 3 | 872 | 69 | |
AY133868.1 | - | 2 - 3 | 803 | 69 | ||
D17582.1 | - | 2 - 3 | 917 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
