Assay Details
Gene Symbol: ERD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124486.3 | NP_568750.1 | 4 - 5 | 1523 | 90 | |
GenBank mRNA | AY035112.1 | - | 4 - 5 | 1209 | 90 | |
AY133868.1 | - | 4 - 5 | 1140 | 90 | ||
D17582.1 | - | 4 - 5 | 1254 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
