Assay Details
Gene Symbol: AT5G51150 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124494.2 | NP_199928.1 | 4 - 5 | 796 | 122 | |
GenBank mRNA | AY045838.1 | - | 4 - 5 | 725 | 122 | |
AY091374.1 | - | 4 - 5 | 675 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
