Assay Details
Gene Symbol: AT5G51150 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124494.2 | NP_199928.1 | 6 - 7 | 1244 | 73 | |
GenBank mRNA | AY045838.1 | - | 6 - 7 | 1173 | 73 | |
AY091374.1 | - | 6 - 7 | 1123 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
