Assay Details
| Gene Symbol: ACD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124508.4 | NP_568756.2 | 10 - 11 | 1476 | 118 | |
| GenBank mRNA | AY362552.1 | - | 10 - 11 | 1433 | 118 | |
More Information
Set Membership: |
|

ACD5
Genomic Map