Assay Details
| Gene Symbol: ACD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344933.1 | NP_001331641.1 | 9 - 10 | 1375 | 113 | |
| NM_124508.4 | NP_568756.2 | 9 - 10 | 1375 | 113 | ||
| GenBank mRNA | AY362552.1 | - | 9 - 10 | 1332 | 113 | |
More Information
Set Membership: |
|

ACD5
Genomic Map