Assay Details
| Gene Symbol: ATIM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345015.1 | NP_001330129.1 | 15 - 16 | 3285 | 82 | |
| NM_124669.3 | NP_200103.1 | 15 - 16 | 3141 | 82 | ||
More Information
Set Membership: |
|

ATIM
Genomic Map