Assay Details
Gene Symbol: ATIM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345015.1 | NP_001330129.1 | 8 - 9 | 1776 | 96 | |
NM_124669.3 | NP_200103.1 | 8 - 9 | 1632 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
