Assay Details
| Gene Symbol: FTSH11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124696.4 | NP_568787.1 | 14 - 15 | 2232 | 64 | |
| GenBank mRNA | AY091086.1 | - | 14 - 15 | 2160 | 64 | |
| AY123027.1 | - | 14 - 15 | 2131 | 64 | ||
More Information
Set Membership: |
|

FTSH11
Genomic Map