Assay Details
| Gene Symbol: FTSH11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124696.4 | NP_568787.1 | 8 - 9 | 1409 | 88 | |
| GenBank mRNA | AY091086.1 | - | 8 - 9 | 1337 | 88 | |
| AY123027.1 | - | 8 - 9 | 1308 | 88 | ||
More Information
Set Membership: |
|

FTSH11
Genomic Map